Proto® TorquePlus™ Sockets, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches (and metric on select models.)
Certifications: ASME B107.5M.
Frequently Bought Together