ProKlenz® NpH Neutral Detergents
Supplier: Steris
Certificates
About this item
ProKlenz® NpH Neutral Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms.This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.
- Efficacy - Neutral formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
- Easy to validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
- Cost savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
- Globally availability - phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
ProKlenz® NpH High performance neutral detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
Frequently Bought Together