VERIFY™ Steam Integrator Strips, Steris
Supplier: Steris
Certificates
About this item
Chemical integrators monitor all critical parameters of steam sterilization (time, temperature and steam quality).
- Color standard for steam on the integrator strip facilitate color comparison
- Vivid color change on the strip provides an immediate verification of exposure to sterilant
Designed for use in standard steam sterilizing processes, including 250 ° F (121 °C) gravity steam, 270 °F (132°C) prevacuum steam and 270°F (132°C) gravity steam.
Frequently Bought Together