HiScreen Capto Q ImpRes Columns, Cytiva
Catalog # 97067-786
Supplier: Cytiva
Product available on GSA Advantage®
Specifications
- Max. Pressure:4 bar [0.4 MPa] (59 psi)
- Capacity:4.7 ml
- Description:HiScreen™ Capto™ Q ImpRes
- Storage:4 to 30 °C, 20% Ethanol
- Bed dimensions:7.7×100 mm
- Flow velocity:100 - 300 cm/h
- Cat. No.:97067-786
- Supplier no.:17547015
Specifications
About this item
HiScreen Capto Q ImpRes columns are prepacked with Capto Q ImpRes ion exchange chromatography medium, and are part of the process development platform available from Cytiva.
- Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification
- 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20 cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
The columns are an excellent choice for method optimization and parameter screening.