BioMerieux Irradiated Tryptic Soy 3P Agar
Catalog # 76106-964
Supplier: Avantor
CAS Number:
Specifications
- Description:Irradiated Tryptic Soy 3P Agar
- Size:100 plates
- Cat. No.:76106-964
Specifications
About this item
Triple Wrapped and Irradiated with LocksureSample Security System.
- Maximized testing efficiency
- One plate for all critical environmentsSimplified inventory management
- Flexible temperature storage nhanced neutralization capability
- To ensure microbial recovery
Culture Media for Cleanrooms and Isolators, 30mL deep fill volume for active air sampling and settling plate applications.