ContactUsLinkComponent
BioMerieux Rapid Dec L. Mono
BioMerieux Rapid Dec L. Mono
  97067-002
 :  
BioMerieux Rapid Dec L. Mono
  97067-002
 :  03700
 :  
  • Description:
    RapiDec L Mono
  • Size:
    20 tests
  • Cat. No.:
    97067-002
  • Supplier no.:
    03700
Rapid Dec Listeria monocytogenes is a rapid micromethod (identification in 6 hours), for the confirmation of typical colonies (blue witha halo), presumed to be L
monocytogenes, which have been isolated on chromID™ Ottaviani Agosti Agar (OAA).