Order Entry
United States
ContactUsLinkComponent
Liquinox® Critical Cleaning Liquid Detergent
Liquinox® Critical Cleaning Liquid Detergent
Catalog # 470201-486
Supplier:  Alconox
Liquinox® Critical Cleaning Liquid Detergent
Catalog # 470201-486
Supplier:  Alconox
Supplier Number:  1201

Specifications

  • Description:
    Liquinox® critical cleaning liquid detergent
  • Size:
    4×3.8 L (1 gal.)
  • Cat. No.:
    470201-486
  • Packaging:
    Bottle
  • pH:
    8.5
  • Supplier no.:
    1201

Specifications

About this item

Liquinox is a concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water.

  • Dilute 1:100
  • Concentrated, phosphate free, biodegradable and readily disposable
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing to give you reliable results and no interfering residues

It can be used to clean healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors.

And it can be used to remove soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemical and solvents.

Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration. Corrosion testing may be advisable.

Cleaning method: Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace.

Certifications: USDA authorized. FDA certified.
Caution: Will foam - not for spray or machine use.