Order Entry
United States
ContactUsLinkComponent
Potassium Phosphate Monobasic/Sodium, Phosphate Dibasic, Buffer, Rigaku
Potassium Phosphate Monobasic/Sodium, Phosphate Dibasic, Buffer, Rigaku
Catalog # 101443-330
Supplier:  Rigaku Reagents
CAS Number:  
undefined
Potassium Phosphate Monobasic/Sodium, Phosphate Dibasic, Buffer, Rigaku
Catalog # 101443-330
Supplier:  Rigaku Reagents
Supplier Number:  1008040
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mL
  • pH buffer type:
    Phosphate
  • Cat. No.:
    101443-330
  • pH:
    8.5
  • Supplier no.:
    EB-0.5KMNDP85-100

Specifications

About this item

Caution: Intended for Research and Development Use Only.