ContactUsLinkComponent
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80, bioMerieux
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80, bioMerieux
Catalog # 95061-514
Supplier:  bioMerieux
CAS Number:  
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80, bioMerieux
Catalog # 95061-514
Supplier:  bioMerieux
Supplier Number:  P2820
CAS Number:  
Specifications
  • Description:
    Sabouraud Dextrose Agar with Lecithin & Polysorbate 80
  • Size:
    500 mL
  • Cat. No.:
    95061-514
  • Supplier no.:
    P2820
Specifications
About this item
Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
  • For isolation and cultivation of fungi from surfaces sanitized with quaternary ammonium compounds
  • 10 x 500 mL
Upon receipt, store plates at 2 – 8 °C away from direct light in an inverted position. Store bottles according to the package label in an upright position. Store plates in original outer wrapping until ready to use. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period.