ContactUsLinkComponent
Sabouraud Dextrose Agar, bioMerieux
Sabouraud Dextrose Agar, bioMerieux
  95061-470
 :  
Sabouraud Dextrose Agar, bioMerieux
  95061-470
 :  P2070
 :  
  • Description:
    Sabouraud Dextrose Agar
  • Size:
    500 mL
  • Cat. No.:
    95061-470
  • Supplier no.:
    P2070
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
  • Media for the isolation and cultivation of fungi
  • 10 x 500 mL
Upon receipt, store plates at 2 – 8 °C away from direct light in an inverted position. Store bottles according to the product label in an upright position. Store plates in original outer wrapping until ready to use. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period.