Tryptic Soy Agar Bottles, Biomerieux
95061-510
:
- Description:Tryptic Soy Agar
- Size:500 mL
- Cat. No.:95061-510
- Supplier no.:P2050
Tryptic Soy Agar is a general-purpose solid medium used for the cultivation and enumeration of a wide variety of fastidious and non-fastidious microorganisms.
- General purpose medium
- 10 x 500 mL
Store according to the package label in an upright position in the dark. Do not freeze. Allow warming of medium to room temperature before inoculation.