ContactUsLinkComponent
Acrylic Light Guide Demonstrator
Acrylic Light Guide Demonstrator
Catalog # 470230-198
Supplier:  Wards
undefined
Acrylic Light Guide Demonstrator
Catalog # 470230-198
Supplier:  Wards
Supplier Number:  PH0587
Specifications
  • Description:
    Acrylic Light Guide Demonstrator
  • Cat. No.:
    470230-198
Specifications
About this item
Acrylic bar for optics experiments.
  • S shaped bar
  • Bottom of bar is white to aid visibility
An 'S' shaped acrylic bar (220 x 20 x 20 mm) which can be used with the Ray box to demonstrate total internal reflection. It helps students to understand the principles of optical communications. The bottom of the bar is white to aid visibility.