ContactUsLinkComponent
Synchronized Digital Ripple Wave Generator
Synchronized Digital Ripple Wave Generator
Catalog # 470230-262
Supplier:  Wards
undefined
Synchronized Digital Ripple Wave Generator
Catalog # 470230-262
Supplier:  Wards
Supplier Number:  PH0769B
Specifications
  • Description:
    Synchronised Digital Ripple Wave Generator
  • Cat. No.:
    470230-262
Specifications
About this item
Stroboscope light synchronizes with ripple generator.
  • For use with ripple tanks in classroom demonstration and physics experiments
  • Synchronizes frequency of stroboscopic flashes with the generated water ripples in the tank
  • Attaches to strobe light and vibration generator via 4 mm leads
  • 120VAC, 50/60Hz
This Digital Ripple Wave Generator is designed for used with ripple tanks. Unit connects to both a stroboscopic light and a vibration generator via 4mm plugs. This is a very unique apparatus which provides a stroboscopic light with pulses to synchronize with a ripple generator. Both these parameters function in fully synchronized fashion to generate the stable and visible ripple pattern. The motion frequency / ripple frequency and their amplitudes are controlled electronically. Frequency range is from 0.5 Hz to 200 Hz. Uses 120 V AC, 50/60 Hz power.