Steel Tuning Forks, Set of 8
Catalog # 470230-246
Supplier: Wards
Specifications
- Description:Steel Tuning Forks, Set of 8
- Cat. No.:470230-246
Specifications
About this item
Steel tuning forks for physics experiments.
- ±5% of stated frequency
Steel Tuning Forks, Set of 8, C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512).