ContactUsLinkComponent
Dynamics Cart 3 Wheel, Set 2 with Hoops
Dynamics Cart 3 Wheel, Set 2 with Hoops
Catalog # 470230-152
Supplier:  Wards
undefined
Dynamics Cart 3 Wheel, Set 2 with Hoops
Catalog # 470230-152
Supplier:  Wards
Supplier Number:  PH0350B
Specifications
  • Description:
    Dynamics Cart, 3 wheel Set 2 with hoops
  • Dimensions:
    300 x 90 mm
  • Cat. No.:
    470230-152
Specifications
About this item
Sold wood and aluminum dynamics cards used for collision experiments.
  • Carts have with low-friction ball-bearing wheels
This pair of cart is constructed from solid wood and aluminum. Fitted with high quality frictionless nylon ball bearing wheels allows for more accurate dynamic measurements. One of the carts is equipped with an exploder piston that is activated by a trigger on the top of the cart. A pair of spring hoops is provided for collision experiments.