Collision in Two Dimensions Apparatus
Catalog # 470230-146
Supplier: Wards
Specifications
- Description:Collision in Two Dimensions Apparatus
- Cat. No.:470230-146
Specifications
About this item
Collison in two dimensions apparatus with accessories included.
- Consists of 25cm one piece track, 3 balls (12 mm dia.), 2 steel, 1 glass, and 1 plumb bob
- Requires a clamp to attach to end of table (not included)
This apparatus consists of a curved track with a base at one end. On the base is a support to hold a ball at the proper height for a center to center collision with a second ball rolling down the track. The track is level so collision occurs only in the horizontal plane, simplifying calculations. Consists of 25 cm one piece track, 3 balls 12 mm diameter, 2 steel, 1 glass and 1 plumb bob.