ContactUsLinkComponent
Force Board Kit
Force Board Kit
Catalog # 470230-136
Supplier:  Wards
undefined
Force Board Kit
Catalog # 470230-136
Supplier:  Wards
Supplier Number:  PH0345
Specifications
  • Description:
    Force Board Kit
  • Cat. No.:
    470230-136
Specifications
About this item
Kit to demonstrate force experiments.
  • Force board grid has a coarse resolution of 5 x 5cm and a fine resolution of 1 × 1 cm
  • Clamp on pulleys are constructed of ABS plastic with low friction ball bearings
  • More than enough masses and string to do any arrangement of vectors and forces
A great free standing device to introduce forces and vectors to your classroom. High contrast grid made it force configurations easy to see from any angle in class
Ordering information: Kit contains: (2) 1.5" Low Friction Single pulleys, (4) S Hooks, (1) Roll of String, (20) 10g Masses, (1) 55 cm x 37.5 cm Backboard with feet