ContactUsLinkComponent
Pulley with Adjustable Clamp
Pulley with Adjustable Clamp
Catalog # 470230-104
Supplier:  Wards
undefined
Pulley with Adjustable Clamp
Catalog # 470230-104
Supplier:  Wards
Supplier Number:  PH0297A
Specifications
  • Description:
    Pulley with AdjusTable Clamp
  • Cat. No.:
    470230-104
Specifications
About this item
Metal pulley with rotatable pulley for experiment set up.
  • Sturdy metal construction throughout
  • Ideal for demonstration of wide variety of mechanics principles
  • 2" (5 cm) dia. sheave
  • Completely rotatable pulley for flexibility in experiment setups
  • Adjustable to 9" height with 1.9" maximum clamp opening
Sturdy, metal clamp pulley can be used to demonstrate a wide variety of mechanics principles. The metal sheave is 5 cm in diameter and can be adjusted from flush with the tabletop to 13 cm above it. Pulley can be rotated to 360º to suit any experimental setup.