Orders LinkContactUsLinkComponent
Volcanic Eruptions
Volcanic Eruptions
Volcanic Eruptions Kit
Catalog # 470231-162
Supplier:   Kemtec
Supplier #:  1-186
Volcanic Eruptions
Catalog # 470231-162
Supplier:  Kemtec
Volcanic Eruptions
Volcanic Eruptions
Volcanic Eruptions Kit
Price
$168.00
Log in for availability
Each
Product Details & Documents
A hands-on volcano model building project complete with safe, simulated lava eruptions.
  • Grades K-8
  • Designed to accommodate 24 students working in groups of four
Learn all about types of volcanoes, parts of a volcano, how volcanoes function, and precious gems volcanoes may produce. Building materials are appropriate for elementary and middle school classrooms. Also investigate underwater volcanoes with the included demonstration.
Ordering information: Baking soda and vinegar required but not included. Kit contains instructor’s Manual, Student Manuals, Student Data Sheet, Plastic Beakers, Gauze, Red dye, Mesh Sheets, Gloves, Duct Tape, Twist Ties, Paint, Brushes, Liquid Dish Detergent, Marbles, and Aluminum Pans.
Specifications
  • Description:
    Volcanic Eruptions Kit