ContactUsLinkComponent
Density, Set Assorted Shapes, Set 14
Density, Set Assorted Shapes, Set 14
Catalog # 470230-054
Supplier:  Wards
undefined
Density, Set Assorted Shapes, Set 14
Catalog # 470230-054
Supplier:  Wards
Supplier Number:  PH0114A
Specifications
  • Description:
    Density Set, Assorted Shapes, Set 14
  • Cat. No.:
    470230-054
Specifications
About this item
Fourteen materials in the form of rectangular blocks.
  • 14 different blocks
  • Kit contains 11 different materials,
  • This kit is intended to familiarize students with appearance, feel, texture and density of these common materials.
  • All measurements are approx. and may vary from kit to kit
Ordering information: Kit consists of 14 blocks: Aluminum Block (2" x 1.5" x 1.125"); Aluminum Rectangle (3.875" x 0.75" x 0.75"); Brass Block (2" x 0.75" x 0.75"); Styrofoam block (2" x 1.5" x 1.125"); Wax Block (2" x 1.5" 1.125"); Stainless Steel Block (2" x 2" x 0.75”); Wood Block (2" x 1.5625" x 1.1875”); Wood Rectangle (3.875" x 0.75" x 0.75"); Soap stone Rectangle (3.875" x 0.75" x 0.75"); Marble Rectangle (3.875" x 0.75" x 0.75"); Glass Rectangle (3.875" x 0.75" x 0.75"); Acrylic Rectangle (3.875" x 0.75" x 0.75"); Steel Block (1.5" x 1.5" x 0.75"); Steel Block (2" x 1.5" x 1.125").