Specifications
- Description:Barlow's Wheel
- Cat. No.:470230-298
Specifications
About this item
Barlow's wheel ideal for demonstrating Lorentz force.
- Barlow's Wheels is an impressive piece of apparatus constituted by a wheel on low friction suspensions
- Wheel does not spin when batteries are installed-This demonstrates the magnetic resistance due to electricity being applied
- 4 AA batteries are required for operation of the Barlow (batteries not included)
Barlow's wheel is an historical demonstration experiment of Lorentz force generated by currents flowing between the polar expansions of a magnet. The item is equipped with neodinium-ironboron magnets that generate a strong magnetic field over the wheel that is therefore set on motion when crossed by currents.
Discover Lorentz force without using mercury. Barlow's wheel is an historical demonstration experiment of Lorentz force generated by currents flowing between the polar expansions of a magnet. Barlow's Wheels is an impressive piece of apparatus constituted by a wheel on low friction suspensions. The item is equipped with neodinium-ironboron magnets that generate a strong magnetic field over the wheel that is therefore set on motion when crossed by currents.