Order Entry
United States
Orders LinkContactUsLinkComponent
 

Plotting Compass, 16 mm

Plotting Compass, 16 mm, 1.3 cm (0.6")
Catalog # 470230-286
Supplier:   Wards
 :  PH0820APK10
Plotting Compass, 16 mm
Catalog # 470230-286
Supplier:  Wards
Plotting Compass, 16 mm
Plotting Compass, 16 mm

Plotting Compass, 16 mm, 1.3 cm (0.6")

Clearance
$4.90
Sale

Valid until 31/12/2026

Log in for availability
Pack of 10
Product Details & Documents

Simple, easy-to-use compasses.

  • Glass face
  • Aluminum case
  • Pack of 10

Basic 16 mm plotting compass with principal points marked. Glass face in aluminum case. These compasses provide a simple, teachable way to demonstrate magnetic fields in class. Simple use and easy readability make it straight-forward for students to map magnetic fields.

Specifications
  • Description:
    Plotting Compass, 16 mm
  • I.D.:
    1.3 cm (0.6")