Order Entry
United States
ContactUsLinkComponent
Density Cubes with Hooks
Density Cubes with Hooks
Catalog # 470230-032
Supplier:  Wards
undefined
Density Cubes with Hooks
Catalog # 470230-032
Supplier:  Wards
Supplier Number:  PH0108PCB

Specifications

  • Description:
    Density Cubes with Hooks
  • Cat. No.:
    470230-032

Specifications

About this item

Density Cubes, Set of 4 Metals with Hooks, 1.26" (32 mm) sides.

  • Brass, lead, zinc, copper, aluminum, and iron cubes
  • 32 mm cubes with hooks

The set consists of four 32mm cubes of the following metals: aluminum, brass, lead, and steel . Perfect for investigating density. Note: this product contains lead. To be used for scientific purposes only.