ContactUsLinkComponent
Color Mixing Demonstrator
Color Mixing Demonstrator
Catalog # 470229-992
Supplier:  Wards
undefined
Color Mixing Demonstrator
Catalog # 470229-992
Supplier:  Wards
Supplier Number:  CMB14L
Specifications
  • Description:
    Color Mixing Demonstrator
  • Cat. No.:
    470229-992
Specifications
About this item
Color Mixing apparatus - 3 LED Lamps that project Red, Blue, Green light.
  • 3 LED Lamps that projects Red, Blue, Green light.
  • Each lamp is flexible and is connected to an individual knob that controls the brightness of the lamp.
  • Light projects approx. 10 - 15 feet.
  • This comes with a 110 v wall plug so no power supply is needed.
  • 10 x 16" base
This labs apparatus - 3 LED Lamps that projects Red, Blue, Green light. Each lamp is flexible and is connected to an individual knob that controls the brightness of the lamp. This comes with a 110 v wall plug so no power supply is needed. The science of light and color is a fun topic which is further amplified by this color mixing table apparatus. The LEDS can be projected onto a piece of paper or wall depending on if you are doing a classroom demonstration or student group bench top experiments. Light projects approx. 10 - 15 feet. 10" x16" base