Order Entry
United States
ContactUsLinkComponent
M9 Salts Broth Media without Glucose and Ammonium Chloride, Teknova
M9 Salts Broth Media without Glucose and Ammonium Chloride, Teknova
Catalog # 550000-020
Supplier:  Avantor
CAS Number:  
M9 Salts Broth Media without Glucose and Ammonium Chloride, Teknova
Catalog # 550000-020
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    M9 Salts Broth Media without Glucose and Ammonium Chloride
  • Size:
    1000 mL
  • Environmentally Preferable:
  • Cat. No.:
    550000-020
  • Volume:
    1 L
  • Supplier no.:
    M8008

Specifications

About this item

M9 is a mimimal, defined growth medium used for cultivating recombinant strains of Escherichia coli.

  • 1000 mL
  • Sterile

Contains 1X M9 Salts, 0.5µg/mL Thiamine, 1 mm Magnesium Sulfate, 0.1 mm Calcium Chloride.

Ambient shipping conditions. Store at room temperature.