Permanent Polypropylene Labels, White, 3" Core, ToughBond®, Brady
The B8425 is a white polypropylene label designed for general purpose use, asset identification, rating plate, and warning/instructional labeling.
- Glossy polypropylene with a permanent acrylic adhesive
- UL Recognized Component and CSA Accepted when printed with the recommended ribbon
- Service temperature of –40 to 194 °F
ToughBond® printable polypropylene labels for thermal transfer printers.
Please Note: This material is UL and CSA approved with its respective ribbon. See the 'Recommended Ribbon Series' for the appropriate ribbon to use. Minimum order of 4 rolls.
: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Frequently Bought Together