Order Entry
United States
ContactUsLinkComponent
MasterPure™ Gram Positive DNA Purification Kit, Biosearch Technologies
MasterPure™ Gram Positive DNA Purification Kit, Biosearch Technologies
Catalog # 76081-754
Supplier:  Lucigen
CAS Number:  
undefined
MasterPure™ Gram Positive DNA Purification Kit, Biosearch Technologies
Catalog # 76081-754
Supplier:  Lucigen
Supplier Number:  MGP04100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Precipitation
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    MasterPure™ Gram Positive DNA Purification Kit, Lucigen / Epicentre
  • Size:
    100 Purifications
  • Description:
    MasterPure™ Gram Positive DNA Purification Kit
  • Cat. No.:
    76081-754

Specifications

About this item

Purify genomic DNA from challenging Gram positive bacteria for a wide variety of molecular biology applications

The MasterPure™ Gram Positive DNA Purification Kit provides all of the reagents needed to purify DNA from Gram positive bacteria

Examples of Gram-positive bacteria tested with this kit include Bacillus subtilis, Listeria monocytogenes, Staphylococcus aureus, Streptococcus mutans, and Lactococcus lactis. The kit effectively purifies high molecular weight genomic DNA which is suitable for PCR analysis and many other molecular biology applications.

Produces clean, high molecular weight genomic DNA ready for PCR and other molecular biology applications such as restriction digestion, cloning, and Southern blotting
Scaleable method for large sample volumes with no wasting of reagents with smaller samples
Complete kit includes lysozyme and does not require any additional purchases
Flexible kit also purifies gDNA from Gram negative bacteria
Robust kit works on a variety of tested species

These bacteria lyse more readily after treatment with Ready-Lyse™ Lysozyme and the Gram-Positive Cell Lysis Solution included in the kit. Ready-Lyse Lysozyme is a stable solution of a recombinant lysozyme from a nonmammalian, nonavian source. It has high specific activity and does not bind DNA.