

Sodium hypochlorite 5 - 6% available chlorine USP
Catalog # 10192-152
Supplier: Hawkins Pharmaceutical Group
CAS Number:
Specifications
- Size:3.8 L (1 gal)
- Cat. No.:10192-152
- Supplier no.:36211
Specifications
Specification Test Results
- Identification A, IR 197:Conforms
- Identification A*:Conforms
- Identification B*:Conforms
- Assay, wt. %:5.0 - 6.0 %
- Appearance:Clear to light yellow
- Phosphate, P04, ppm*:≤5 ppm
- Calcium, Ca, ppm*:≤10 ppm
Specification Test Results
About this item
:
:74.44 g/mol
UN:1791
ADR:8,III
:GHS05
:GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120