Buffer Solution, Butterfields Phosphate Buffer, pH 7.2, Moltox
1000 mL, Bottle, 7.2, Phosphate
Supplier: MOLTOX Molecular Toxicology
Supplier #: 26-622
Buffer Solution, Butterfields Phosphate Buffer, pH 7.2, Moltox
Catalog # 101106-840
Supplier: MOLTOX Molecular Toxicology
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Size:1000 mL
- pH buffer type:Phosphate
- pH:7.2
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



