Order Entry
United States
ContactUsLinkComponent
Chromium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Chromium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Catalog # 100515-431
Supplier:  Spex Certiprep
CAS Number:  
Chromium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Catalog # 100515-431
Supplier:  Spex Certiprep
Supplier Number:  SPEC-CR6
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Plastic Bottle
  • Size:
    125 ml
  • Cat. No.:
    100515-431
  • Element:
    Chromium
  • Supplier no.:
    SPECCR6

Specifications

About this item

  • Traceable to NIST
  • Uncertainty of standards is reported on the Certificate of Analysis
  • Manufactured by experienced chemists to ensure highest quality standards
Caution: 1000 µg/mL = 1000 ppm