Ward's® Essentials Design a Coated Pill
470231-274
- Description:Design a Coated Pill
- Cat. No.:470231-274
Explore Biomedical Engineering.
- Illustrates dissolving, diffusion, and pH
- Includes real-world problem solving
- Aligned with NGSS 5-PS1-3, 3-5-ETS1-1, MS-ETS1-1, MS-ETS1-2, MS-ETS1-3, MS-ETS1-4
Students use provided materials to formulate a coating for a pill that will withstand the harsh environment of the stomach. Students will test and modify their designs to determine which coating protects the best.
: Kit includes materials for 15 groups to create a protective coating, and a teacher's guide.