Order Entry
United States
ContactUsLinkComponent
Complex Cyanide Standard, Agilent Technologies
Complex Cyanide Standard, Agilent Technologies
Catalog # ULICC-009
Supplier:  Ultra Scientific
Small Business Enterprise
CAS Number:  
undefined
Complex Cyanide Standard, Agilent Technologies
Catalog # ULICC-009
Supplier:  Ultra Scientific
Supplier Number:  ICC-009
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Description:
    Complex Cyanide Standard
  • Size:
    125 ml
  • Cat. No.:
    ULICC-009
  • Contents:
    K₃[Fe(CN)₆] in 0.1% NaOH
  • Supplier no.:
    ICC-009

Specifications

About this item