Orders LinkContactUsLinkComponent
LS Columns, Miltenyl Biotec
LS Columns, Miltenyl Biotec
LS Columns and plungers
Catalog # MSPP-130042401
Marketsource
Supplier:   Miltenyi Biotec
Supplier #:  130-042-401
LS Columns, Miltenyl Biotec
Catalog # MSPP-130042401
Supplier:  Miltenyi Biotec
Marketsource
LS Columns, Miltenyl Biotec
LS Columns and plungers
Price
$600.00
Log in for availability
Pack of 25
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
LS Columns are designed for positive selection of cells. They are also suitable for the depletion of strongly magnetically labeled cells.
LS Columns are designed for positive selection of cells
The matrix of the LS Columns is composed of ferromagnetic spheres, which are covered with a cell-friendly coating allowing fast and gentle separation of cells. It can be used to separate material less than 30 μm in size.
LS Columns can be used with the following separators MidiMACS™, QuadroMACS™, VarioMACS™ in combination with an LS column adapter, SuperMACS™ II and MultiMACS Cell24 Separator Plus in combination with the single column adapter.
They are also suitable for the depletion of strongly magnetically labeled cells.
Packaging: The columns are individually packed in a sterile way.
Specifications
  • Description:
    LS Columns and plungers