ContactUsLinkComponent
Immersion Oil, Type A, Low Viscosity, Hardy Diagnostics
Immersion Oil, Type A, Low Viscosity, Hardy Diagnostics
Catalog # 76076-690
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Immersion Oil, Type A, Low Viscosity, Hardy Diagnostics
Catalog # 76076-690
Supplier:  Hardy Diagnostics
Supplier Number:  Z95
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Immersion Oil, Type A, Low Viscosity
  • Size:
    20 mL
  • Cat. No.:
    76076-690
Specifications
About this item
Used to assist in microscopic examination at high power (1,000 X lens) by increasing the resolving power
Type A is a low viscosity Immersion Oil.