E-Z Press™ hydraulic lab presses feature built in polycarbonate safety shields with solid brass hardware and unique lightweight ergonomic design.
- Unibody construction
- Comprises adjustable overhead lead screw
- Access ports for vacuum and electrical lines
The 15 and 20 ton E-Z Press™ will handle a variety of lab applications, including pressing KBr pellets for FTIR spectroscopy and briquetting samples for XRF spectroscopy and can also be used for making thin films with ICL’s precision thickness film maker and heated platens
The bottom platen of the 12 ton E-Z Press™ automatically centers ICL’s 13 mm Macro-Micro die. The bottom platen of the 15 and 20 ton E-Z Press™ has a 4" diameter depression and optional donut shaped inserts are available to automatically center ICL’s 1.3 cm, 2 cm, 3.2 cm, 3.5 cm and 4 cm die sets.
The Model 30 and Model 40 features the same patented lightweight and ergonomic unibody design as the 12 ton and 20 ton models. To make what would normally be huge presses as light and elegant as the lower capacity designs,they have the finest quality low profile hydraulic cylinders available. Oversized hydraulic cylinders make them as easy as possible to pump to full load. Both presses have an award winning 2 speed hand pump that reduces the number of required strokes by up to 78% and also reduces jacking effort. These high tonnage E-Z presses are ideal for lab applications, such as briquetting samples for XRF spectroscopy, because of their ease of use, small footprint, light weight construction and built in safety features.
There are die centering depressions in the round bottom platens. Each press comes with a standard stainless steel filling disc to flatten the platen.
- Catalog No:
- 100215-578
- 100215-576
- 100216-100
- 100216-080
- 100215-378
- 100215-574
- Model:
- 0012-6629
- 0012-6549
- 0012-5859
- 0012-6306
- 0012-6630
- 0012-6266
- W×D×H:
- 20×15×21"
- 20×15×21"
- 14×10×15.5"
- 18×12×17.5"
- 20×15×21"
- 20×15×21"
- Weight:
- 126 lbs.
- 129 lbs.
- 50 lbs.
- 90 lbs.
- 126 lbs.
- 126 lbs.