Order Entry
United States
ContactUsLinkComponent
FosmidMAX™ DNA Purification Kit, Biosearch Technologies
FosmidMAX™ DNA Purification Kit, Biosearch Technologies
Catalog # 76085-634
Supplier:  Avantor
CAS Number:  
FosmidMAX™ DNA Purification Kit, Biosearch Technologies
Catalog # 76085-634
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Larger vectors (Cosmid, BAC, PAC, Fosmid)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    FosmidMAX™ DNA Purification Kit, Lucigen/Epicentre
  • Size:
    100 x 1.5 mL
    10 x 40 mL
    5 x 100 mL
  • Environmentally Preferable:
  • Description:
    FosmidMAX™ DNA Purification Kit
  • Cat. No.:
    76085-634

Specifications

About this item

Easy, reliable isolation of high-quality fosmid DNA

The FosmidMAX™ DNA Purification Kit was developed for easy, reliable isolation of high-quality fosmid DNA

The FosmidMAX kit is ideal for the purification of induced fosmids produced by the CopyControl™ Fosmid Library Production Kit. The CopyControl cloning technology enables the user to grow the fosmid clones at single copy to ensure insert stability and cloning of toxic gene products and then induce the clones to high-copy number for high yields of DNA. Using the FosmidMAX Kit, up to 4 µg of fosmid DNA can be isolated from 1.5 ml of an induced clone.

Isolate up to 0.6, 15, or 25 µg of fosmid DNA from 1.5-, 40-, or 100-ml cultures of a single-copy fosmid, respectively, with an easy to use protocol
Maximize yields because the FosmidMAX kit does not use columns or binding resins that reduce yields
Reduce costs by using only the reagents you need based on the amount of DNA being purified
Purify DNA from single-copy fosmid clones, induced CopyControl fosmid and cosmid clones
Reduce waste and use of hazardous reagents because no columns or toxic organic solvents are used

The scalable protocol is based on a modified alkaline-lysis procedure that typically yields 0.6-25 µg of fosmid DNA from 1.5 to 100 ml of a single-copy fosmid culture. Selective precipitation steps and the incorporation of the Epicentre RiboShredder™ RNase Blend effectively remove contaminants that degrade DNA and interfere with downstream applications.