Order Entry
United States
ContactUsLinkComponent
 
Tris EDTA Buffer pH 9 (20X), Concentrate, 100 ml, Genemed
  75814-850
 :  Avantor
 :  
Tris EDTA Buffer pH 9 (20X), Concentrate, 100 ml, Genemed
  75814-850
 :  Avantor
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Size:
    100 mL
  • Form:
    Liquid
  • Storage Temperature:
    4°C
  • Brand Name:
    Genemed
  • Shipping Temperature:
    4°C
  • Cat. No.:
    75814-850
  • Supplier no.:
    10-0037

 

 

For In Vitro Diagnostic Use.
This product is intended to be used for the heat induced epitope (or antigen) retrieval (HIER) of formalin-fixed, paraffin-embedded (FFPE) tissues prior to immunohistochemical (IHC) staining.
One bottle of Tris EDTA Buffer, pH 9 containing 200 mM Tris and 20 mM EDTA.

Formalin fixation forms protein cross-links that mask the antigenic sites in tissue specimens, thereby giving weak or false negative staining for IHC detection of certain proteins. Tris EDTA Buffer, pH 9 is designed to break the protein cross-links, thus unmasking the antigens and epitopes in FFPE tissue sections and enhancing staining intensity of many antibodies.

For use after de-parafinnizing and rehydrating slides. If necessary, block endogenous peroxidase activity before HIER step.
1. Make 1X Tris EDTA Buffer using 1 part 20X Tris EDTA Buffer and 19 parts Reagent Water.
2. Wash slides in 3 changes of 1X PBS or reagent water to remove alcohol / peroxidase block.
3. Place slides in appropriate sized slide container and fill with 1X Tris EDTA Buffer. Make sure section is immersed.

4. Incubate for 20-40 minutes at temperature ≥ 95°C.

Note: Optimal incubation time in variable heating source should be determined by user.