Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant WNT3A (from E. coli)
 :  Avantor
 
undefined
Human Recombinant WNT3A (from E. coli)
 :  Avantor

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Gel Pack
  • Gene ID:
    P56704
  • Protein Synonyms:
    WNT3A|Protein Wnt-3a
  • Protein/Peptide Name:
    WNT3A
  • Purity:
    > 85%
  • Molecular Weight:
    37.5 kDa
  • Formulation:
    Liquid

 

Frequently Bought Together