MasterPure™ Bulk Reagents, Reagents for Nucleic Acid Purification, Biosearch Technologies
Supplier: Lucigen
Certificates
About this item
Save by purchasing bulk quantities of the MasterPure™ buffers and reagents
- Save: Reduce costs by purchasing these important reagents in bulk quantities
- Scale-up: Use these large volumes of reagents to perform large-scale MasterPure™ purifications
- Adapt: Use these reagents whenever your experiments require these high quality reagents
Proteinase K: Useful for multiple applications outside of the MasterPure™ Kits including (i) isolation of plasmids and high molecular weight gDNA, purification of intact RNA, and inactivation of RNases, DNases and other enzymes in molecular biology reactions
Red Cell Lysis Solution: Selectively lyse red blood cells while leaving white blood cells intact
Tissue and Cell Lysis Solution: Use in combination with Proteinase K to lyse cells and tissues in various samples such as saliva, semen, mammalian cells, and gram negative bacteria
Certifications: For research use only. Not for human or diagnostic use.
Frequently Bought Together