Endonuclease
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Remove all types of RNA and DNA with this robust, flexible nuclease
- Non-specific: Digests all types of DNA and RNA including single- or double-stranded linear, circular, and supercoiled
- Flexible: Active within a broad range of conditions normally used in protein purification
- Customizable: Contact us for bulk or high concentration enzyme
OmniCleave™ Endonuclease is a highly purified enzyme from a recombinant E
Applications include:
Removal of nucleic acids from cell lysates (reduction of viscosity) for improved handling and yield of protein preparations
Removal of trace contamination by nucleic acids in protein preparations
Removal of host DNA from phage preparations
Improved electrophoretic and chromatographic separation of proteins isolated from whole-cell lysates
coli strain that degrades single- and double-stranded DNA and RNA to di-, tri-, and tetranucleotides. OmniCleave Endonuclease has the same substrate specificity and yields the same products as Benzonase®, an enzyme derived from Serratia marcescens.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a dilution buffer.
Documents
Omnicleave-Endonuclease
Manual
Specifications
Specifications
- Size:50000 units
- Cat. No.:76081-706
- Concentration:200 units/μl
- Enzyme Name:Endonuclease
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



