Order Entry
United States
Orders LinkContactUsLinkComponent
RNase A

RNase A

2 ml
Catalog # 76081-698
Supplier:   Lucigen
Supplier #:  MRNA092
RNase A
Catalog # 76081-698
Supplier:  Lucigen
CAS Number:  
RNase A
RNase A

2 ml

CAS Number:  
Price
$228.22
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Remove unwanted single-stranded RNA from multiple sample types or reactions

  • Quick: Rapidly degrade single-stranded RNA with this highly effective endoribonuclease
  • Robust: RNase A is a durable enzyme and resistant to many harsh treatments (e.g. boiling)

Ribonuclease A (RNase A) is an endoribonuclease that cleaves single-stranded RNA at the 3´ end of pyrimidine residues, forming oligoribonucleotides having 3´-terminal pyrimidine-3´-phosphates

Applications include:
Removal of RNA from DNA preparations
Removal of unhybridized regions of RNA from DNA-RNA or RNA-RNA hybrids

Pyrimidine-3´-monophosphates are also released by RNase A cleavage of adjacent pyrimidine nucleotides. Modified RNA containing pyrimidine-2´-fluoro-dNMPs, such as DuraScript® RNA made by in vitro transcription using the DuraScribe® T7 & SP6 Transcription Kits is completely resistant to cleavage by RNase A.

Certifications: For research use only. Not for human or diagnostic use.

Specifications

Specifications

  • Size:
    2 ml
  • Cat. No.:
    76081-698
  • Concentration:
    5 µg/µl
  • Enzyme Name:
    RNase A

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.