Order Entry
United States
ContactUsLinkComponent
E. coli Exonuclease III
E. coli Exonuclease III
Catalog # 76085-632
Supplier:  Lucigen
CAS Number:  
undefined
E. coli Exonuclease III
Catalog # 76085-632
Supplier:  Lucigen
Supplier Number:  EX4425K
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    E. coli
  • Size:
    25000 units
  • Enzyme Name:
    Exonuclease III
  • Concentration:
    200 units/μl
  • Cat. No.:
    76085-632

Specifications

About this item

Digest duplex DNA in a 3´ to 5´ direction from a nick, blunt end, or 3´-recessed end, producing stretches of ssDNA

  • Produce stretches of ssDNA in dsDNA templates containing nicks, blunt ends or recessed 3' ends
  • Protect protruding 3' ends becuase this exonuclease will not start digestion at protruding 3' overhangs

Exonuclease III digests duplex DNA in a 3´ to 5´ direction from a nick, a blunt end, or 3´-recessed end, producing stretches of ssDNA on the opposite strand. Under defined reaction conditions, DNA degradation by Exonuclease III proceeds at a uniform rate yielding predictable and reproducible digestion results.

Because the rate of exonucleolytic excision of deoxyribonucleotides by Exonuclease III is dependent upon reaction factors including temperature, ionic strength, template sequence, and enzyme-to-DNA ratios, each template must be optimized using sample digestions to achieve the desired excision rate.

Exonuclease III is not active on 3´-protruding ends of four bases or more in length, ssDNA, or on thioester-linked nucleotides. The enzyme also has intrinsic RNase H, 3´-DNA phosphatase, and apurinic DNA endonuclease activities.

Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer.