Order Entry
United States
ContactUsLinkComponent
 
E. coli Exonuclease I
  76081-722
 :  Lucigen
 :  
undefined
E. coli Exonuclease I
  76081-722
 :  Lucigen
 :  X40520K
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Species:
    E. coli
  • Size:
    20000 units
  • Enzyme Name:
    Exonuclease I
  • Concentration:
    20 units/μl
  • Cat. No.:
    76081-722

 

 

Digest single-stranded DNA from the 3' to 5' end, but not double-stranded or dsDNA with short 3' overhangs

  • Specific: Processively digest ssDNA in a 3´ to 5´ direction without harming dsDNA - short 3´ overhangs in DNA are not digested
  • Flexible: Active under a wide variety of buffer conditions and can be added directly into most reaction mixes
  • Heat Inactivated: Destroy the enzyme activity by incubating at 80°C for 15 minutes

Exonuclease I digests ssDNA in a 3´ to 5´ direction,1-3 but does not digest dsDNA

Applications include:
Removal of residual ssDNA, including oligos, from reaction mixes
Removal of ssDNA from nucleic acid mixtures

Although it requires the presence of magnesium and a free 3´-hydroxyl terminus for activity, it is active under a wide variety of buffer conditions and can be added directly into most reaction mixes. Exonuclease I can be heat-inactivated by incubation at 80°C for 15 minutes.

 : For research use only. Not for human or diagnostic use.