Order Entry
United States
ContactUsLinkComponent
T4 Polynucleotide Kinase, Lucigen
T4 Polynucleotide Kinase, Lucigen
Catalog # 76081-708
Supplier:  Lucigen
CAS Number:  
undefined
T4 Polynucleotide Kinase, Lucigen
Catalog # 76081-708
Supplier:  Lucigen
Supplier Number:  P0503K
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    3000 units
  • Enzyme Name:
    T4 Polynucleotide Kinase
  • Concentration:
    10 units/μl
  • Cat. No.:
    76081-708

Specifications

About this item

Phosphorylate 5' hydroxy ends of ssDNA and dsDNA, RNA, and nucleoside 3´ monophosphates

  • Robust: Highly active enzyme that readily phosphorylates 5' hydroxl ends of nucleic acids
  • Flexible: Use newly phosphorylated nucleic acids in a variety of applications such as cloning, next gen sequencing library prep, and preparation of labeled nucleic acids

T4 Polynucleotide Kinase (PNK) catalyzes the transfer of the γ-phosphate from ATP to the 5´ hydroxyl of ssDNA and dsDNA, RNA, and nucleoside 3´ monophosphates

Applications include:
Labeling of 5´ termini of DNA and RNA with 32P or 33P for DNA sequencing, blot-hybridization experiments, or transcript mapping using Mung Bean Nuclease, S1 nuclease, or other nucleases
Phosphorylation of oligonucleotide linkers and other DNA or RNA molecules prior to ligation, or for use in ligation amplification reactions with Ampligase® Thermostable DNA Ligase
Preparation of labeled DNA or RNA molecular weight markers for gel electrophoresis and chromatography

The enzyme also removes the 3´ phosphate from 3´-phosphoryl polynucleotides, deoxyribonucleoside 3´ monophosphates, and deoxyribonucleoside-3´,5´-diphosphates to form a 3´-hydroxyl group.

Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer without ATP. ATP must be purchased separately.