Order Entry
United States
Orders LinkContactUsLinkComponent
Poly (A) Polymerase

Poly (A) Polymerase

50 reactions
Catalog # 76081-710
Supplier:   Lucigen
Supplier #:  PAP5104H
Poly (A) Polymerase
Catalog # 76081-710
Supplier:  Lucigen
CAS Number:  
Poly (A) Polymerase
Poly (A) Polymerase

50 reactions

CAS Number:  
Price
$555.57
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Polyadenylate 3' ends of RNA molecules for various downstream applications

  • Quality Kit: High enzymatic purity, specificity, and activity
  • Flexible: Use polyadenylated RNAs in a variety of applications such as stabilizing in vitro transcribed RNA and adding primer (oligo-dT) binding sites to any RNA for first-strand cDNA synthesis
  • Fast and Easy: Simple protocol produces polyadenylated RNA molecules quickly

Poly(A) Polymerase uses ATP as a substrate for template-independent addition of adenosine monophosphate to the 3´-hydroxyl termini of RNA molecules. The Poly(A) Polymerase Tailing Kit provides the enzyme and other reagents for quickly and easily adding a poly(A) tail to the 3´ end of any RNA

Applications include:
Addition of a poly(A) tail to RNA synthesized in vitro in order to increase the stability of the RNA and enhance its ability to be translated in vivo after transfection or microinjection into eukaryotic cells
Addition of a poly(A) tail to an RNA molecule or a mixture of RNA molecules in order to provide a priming site for synthesis of first-strand cDNA using a primer with poly(dT) on its 3´ end
Synthesis of polyadenylated RNA for nucleic acid amplification methods or gene expression studies
3´-end labeling of RNA with radioactive A residues
Quantifying mRNA

Poly(A) Polymerase is encoded by an E. coli gene that has been cloned into a plasmid and overexpressed in an E. coli strain.

Certifications: For research use only. Not for human or diagnostic use.

Ordering information: Contents include Poly(A) Polymerase, 10X Reaction Buffer, 10 mM ATP, and Sterile RNase-Free Water.

Specifications

Specifications

  • Size:
    50 reactions
  • Cat. No.:
    76081-710
  • Enzyme Name:
    Poly (A) Polymerase

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.