Orders LinkContactUsLinkComponent
ToughCut® Fine Scissors, Fine Science Tools
Marketsource
ToughCut® Fine Scissors, Fine Science Tools
Marketsource
Certificates
ToughCut® Fine Scissors, Fine Science Tools
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Product Family Options
Product Information
  • Length
    Material
    Blade type
    Curvature
    Availability
    Price
  • MSPP-1405809
    Cutting edge 22 mm
    Cutting edge 22 mm
    9 cm
    Stainless steel
    Sharp/Sharp, Serrated
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $156.92
    $156.92
    Each
       $156.92
    Supplier Number: 14058-09
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1405811
    Cutting edge 25 mm
    Cutting edge 25 mm
    11.5 cm
    Stainless steel
    Sharp/Sharp, Serrated
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $149.23
    $149.23
    Each
       $149.23
    Supplier Number: 14058-11
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1405911
    Cutting edge 25 mm
    Cutting edge 25 mm
    11.5 cm
    Stainless steel
    Sharp/Sharp, Serrated
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $175.38
    $175.38
    Each
       $175.38
    Supplier Number: 14059-11
    Specifications:
    Marketsource
    Marketsource