Order Entry
United States
ContactUsLinkComponent
FFAU, [6-3H] ≥97% (by HPLC), MORpure™
Irritant
FFAU, [6-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-792
CAS Number:  
undefined
Irritant
FFAU, [6-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-792
Supplier Number:  MT853-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101973-792
  • Supplier no.:
    MT853-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:340128-68-9