Order Entry
United States
ContactUsLinkComponent
Puromycin dihydrochloride, [8-3H(N)] ≥97% (by HPLC), MORpure™
Irritant
Puromycin dihydrochloride, [8-3H(N)] ≥97% (by HPLC), MORpure™
Catalog # 101973-614
CAS Number:  
undefined
Irritant
Puromycin dihydrochloride, [8-3H(N)] ≥97% (by HPLC), MORpure™
Catalog # 101973-614
Supplier Number:  MT785-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101973-614
  • Supplier no.:
    MT785-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:73430-35-0