Order Entry
United States
ContactUsLinkComponent
5-Trifluoromethyl-2'-deoxyuridine, [2'-3H] ≥97% (by HPLC), MORpure™
Irritant
5-Trifluoromethyl-2'-deoxyuridine, [2'-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-550
CAS Number:  
undefined
Irritant
5-Trifluoromethyl-2'-deoxyuridine, [2'-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-550
Supplier Number:  MT751-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101973-550
  • Supplier no.:
    MT751-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:339527-39-8