Order Entry
United States
ContactUsLinkComponent
5-Bromouracil, [6-3H] ≥97% (by HPLC), MORpure™
Irritant
5-Bromouracil, [6-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-244
CAS Number:  
undefined
Irritant
5-Bromouracil, [6-3H] ≥97% (by HPLC), MORpure™
Catalog # 101973-244
Supplier Number:  MT629-250UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    250 µCi
  • Cat. No.:
    101973-244
  • Supplier no.:
    MT629-250UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:33548-49-1